4-Fluorophenethyl bromide, 97%
4-Fluorophenethyl bromide, 97%
4-Fluorophenethyl bromide, 97%
4-Fluorophenethyl bromide, 97%
Thermo Scientific Chemicals
4-Fluorophenethyl bromide, 97%
CAS: 332-42-3 | C8H8BrF | 203.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3980500505 gGlass bottle
Catalog number ACR398050050
Price (MYR)
730.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
EA
Add to cart
Specifications
Chemical Name or Material4-Fluorophenethyl bromide
CAS332-42-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0