Pyridine, for HPLC
Pyridine, for HPLC
Pyridine, for HPLC
Thermo Scientific Chemicals
Pyridine, for HPLC
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3968000101 LGlass Bottle
Catalog number ACR396800010
Price (MYR)
1,696.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,696.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
Name NoteFor HPLC
Absorbance0.02 max. (1 cm cell vs HPLC-grade water) at 345nm, 0.01 max. (1 cm cell vs HPLC-grade water) at 400nm, 0.05 max. (1cm cell vs HPLC-grade water) at 340nm
CAS110-86-1
View more
For HPLC
RUO – Research Use Only