Hydrogen chloride, ca. 0.5M solution in methanol
Hydrogen chloride, ca. 0.5M solution in methanol
Hydrogen chloride, ca. 0.5M solution in methanol
Hydrogen chloride, ca. 0.5M solution in methanol
Thermo Scientific Chemicals
Hydrogen chloride, ca. 0.5M solution in methanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR391748000
Price (MYR)
1,226.00
EA
Add to cart
Quantity:
800 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,226.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 67-56-1
Specifications
Chemical Name or MaterialHydrochloric acid
ColorColorless
Name Note0.5M solution in methanol
CAS7647-01-0
Concentration0.4 to 0.6M