N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5+%, for analysis
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Catalog number ACR390800010
Price (MYR)
707.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
707.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
Name NoteFor analysis
CAS127-19-5
CAS Min %99.5
View more
For analysis
RUO – Research Use Only