4-Bromo-1-butanol, 85+%
4-Bromo-1-butanol, 85+%
4-Bromo-1-butanol, 85+%
4-Bromo-1-butanol, 85+%
Thermo Scientific Chemicals
4-Bromo-1-butanol, 85+%
CAS: 33036-62-3 | C4H9BrO | 153.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3900100505 gGlass bottle
Catalog number ACR390010050
Price (MYR)
1,203.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,203.00
EA
Add to cart
Specifications
Chemical Name or Material4-Bromo-1-butanol
CAS33036-62-3
CAS Min %85.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0