cis-2-Hexene, 95%, Thermo Scientific™
cis-2-Hexene, 95%, Thermo Scientific™
cis-2-Hexene, 95%, Thermo Scientific™
cis-2-Hexene, 95%, Thermo Scientific™
cis-2-Hexene, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR3899400505 mLGlass bottle
Catalog number ACR389940050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialcis-2-Hexene
CAS7688-21-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only