Calcium phosphate, dibasic dihydrate, 98+%, extra pure
CAS: 7789-77-7 | CaHO4P·2H2O | 172.09 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR389830010
1 kg
Plastic bottle
Catalog number ACR389830010
Price (MYR)
575.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
575.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium phosphate, dibasic dihydrate
CAS7789-77-7
ColorWhite
Health Hazard 1GHS Signal Word: Warning
Physical FormPowder