2-Butanone, 99.7%, for HPLC
2-Butanone, 99.7%, for HPLC
2-Butanone, 99.7%, for HPLC
2-Butanone, 99.7%, for HPLC
Thermo Scientific Chemicals
2-Butanone, 99.7%, for HPLC
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3895700101 LGlass Bottle
Catalog number ACR389570010
Price (MYR)
689.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
689.00
EA
Add to cart
Specifications
Chemical Name or Material2-Butanone
Melting Point-87.0°C
Absorbance0.009 max. (1 cm cell vs water) at 350nm, 0.009 max. (1 cm cell vs water) at 450nm, 0.097 max. (1 cm cell vs water) at 340nm, 0.699 max. (1 cm cell vs water) at 330nm, 1 max. (1 cm cell vs water) at 329nm
CAS78-93-3
Health Hazard 1Danger
View more
For HPLC
RUO – Research Use Only