2-Butanone, ACS reagent
2-Butanone, ACS reagent
2-Butanone, ACS reagent
2-Butanone, ACS reagent
Thermo Scientific Chemicals
2-Butanone, ACS reagent
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3894300101 LGlass Bottle
Catalog number ACR389430010
Price (MYR)
575.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
575.00
EA
Add to cart
Specifications
Chemical Name or Material2-Butanone
Melting Point-87.0°C
CAS78-93-3
ColorGolden to Orange
Health Hazard 1Danger