Hydrochloric acid, pure, ca. 32% solution in water
Hydrochloric acid, pure, ca. 32% solution in water
Hydrochloric acid, pure, ca. 32% solution in water
Hydrochloric acid, pure, ca. 32% solution in water
Thermo Scientific Chemicals
Hydrochloric acid, pure, ca. 32% solution in water
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3893100252.5 LGlass bottle
Catalog number ACR389310025
Price (MYR)
572.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
572.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric acid
ColorColorless
CAS7647-01-0
Melting Point-35.0°C
Boiling Point57.0°C