n-Hexane, for analysis, contains appr. 95% n-Hexane
n-Hexane, for analysis, contains appr. 95% n-Hexane
n-Hexane, for analysis, contains appr. 95% n-Hexane
n-Hexane, for analysis, contains appr. 95% n-Hexane
Thermo Scientific Chemicals
n-Hexane, for analysis, contains appr. 95% n-Hexane
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3838000505 LFluorinated HDPE Bottle
Catalog number ACR383800050
Price (MYR)
1,084.00
LT
Add to cart
Quantity:
5 L
Packaging:
Fluorinated HDPE Bottle
Request bulk or custom format
Price (MYR)
1,084.00
LT
Add to cart
Specifications
Chemical Name or Materialn-Hexane
CAS110-54-3
Health Hazard 1Danger
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.
View more
For analysis
RUO – Research Use Only