Benzaldehyde, 99.5+%, pure, redistilled, AcroSeal™
Product Image
Thermo Scientific Chemicals
Benzaldehyde, 99.5+%, pure, redistilled, AcroSeal™
CAS: 100-52-7 | C7H6O | 106.12 g/mol
Have Questions?
Catalog NumberQuantity
ACR378361000100 mL
Catalog number ACR378361000
Price (MYR)
552.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
552.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzaldehyde
CAS100-52-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.