Ethyl isothiocyanate, 96%
Ethyl isothiocyanate, 96%
Ethyl isothiocyanate, 96%
Ethyl isothiocyanate, 96%
Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%
CAS: 542-85-8 | C3H5NS | 87.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR376861000
Price (MYR)
4,177.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,177.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl isothiocyanate
CAS542-85-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.