2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
Thermo Scientific Chemicals
2,4-Dimethoxybenzylamine, 98%
CAS: 20781-20-8 | C9H13NO2 | 167.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3699500505 mLGlass bottle
Catalog number ACR369950050
Price (MYR)
490.00
EA
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
EA
Add to cart
Specifications
Chemical Name or Material2, 4-Dimethoxybenzylamine
CAS20781-20-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0