Check Order Status
Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™
Element Search
Structure Search
Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™
Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736
Have Questions?
Catalog Number
Quantity
Packaging
ACR369722500
250 g
Plastic bottle
Catalog number
ACR369722500
Price (MYR)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Amberlite™ IR120
Physical Form
Beads
CAS
78922-04-0
Particle Size
Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0%
Packaging
Plastic bottle
View more
Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736
Ion exchange resin
RUO – Research Use Only