alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP
alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP
alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP
alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP
Thermo Scientific Chemicals
alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP
alpha-D-Lactose monohydrate, synonym lactose, CAS # 5989-81-1, is the hydrate form of the disaccharide lactose. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3688900101 kgPlastic bottle
Catalog number ACR368890010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Materialα-D-Lactose monohydrate
CAS5989-81-1
PackagingPlastic bottle
Molecular FormulaC12H24O12
Quantity1 kg
View more
General Description
  • Lactose is a disaccharide composed of glucose and galactose
  • Lactose is the major sugar found in dairy products such as milk and cheese
  • alpha-D-Lactose monohydrate is used as a carrier and stabilizer of products in the pharmaceutical and food industries
Applications
  • Lactose has been used in vitro to interact with carbohydrate recognition domains on a recombinant form of LGALS9, thus blocking downstream functions of the protein
  • It has been used as a control to study sugar dependency in Drosophila, which do not digest lactose
  • As a common component of pharmaceutical tablets, alpha-lactose has been used in laboratory settings to imitate drug administration
RUO – Research Use Only
General References:
  1. Walker, G.; Holland, C.R.; Ahmad, M.; Duncan, Q.M. Influence of process parameters on fluidised hot-melt granulation and tablet pressing of pharmaceutical powders. Chem. Eng. Sci.. 2005, 60 (14), 3867-3877.
  2. Montanuy, H.; Martínez-Barriocanal, A.; Casado, J.A. et al. (14 more authors) (2020)
  3. Gefitinib and afatinib show potential efficacy for Fanconi anemia-related head and neck
  4. cancer. Clinical Cancer Research, 26 (12). pp. 3044-3057. ISSN 1078-0432
  5. Wiersma, V.R.; de Bruyn, M.; Wei, Y.; van Ginkel, R.J.; Hirashima, M.; Niki, T.; Nishi, N.; Zhou, J.; Pouwels, S.D.; Samplonius, D.F.; Nijman, H.W.; Eggleton, P.; Helfrich, W.; Bremer, E. The epithelial polarity regulator LGALS9/galectin-9 induces fatal frustrated autophagy in KRAS mutant colon carcinoma that depends on elevated basal autophagic flux. Autophagy. 2015, 11(8), 1373-88.
  6. Umezaki, Y.; Hayley, S.E.; Chu, M.L.; Seo, H.W.; Shah, P.; Hamada, F.N. Feeding-State-Dependent Modulation of Temperature Preference Requires Insulin Signaling in Drosophila Warm-Sensing Neurons. Curr Biol. 2018, 28(5), 779-787.e3.