Ammonia, 2M solution in methanol, AcroSeal™
Ammonia, 2M solution in methanol, AcroSeal™
Ammonia, 2M solution in methanol, AcroSeal™
Ammonia, 2M solution in methanol, AcroSeal™
Ammonia, 2M solution in methanol, AcroSeal™
Thermo Scientific Chemicals
Ammonia, 2M solution in methanol, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR368391000100 mLAcroSeal™ Glass Bottle
Catalog number ACR368391000
Price (MYR)
646.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
646.00
EA
Add to cart
Chemical Identifiers
CAS7664-41-7, 67-56-1
Specifications
Chemical Name or MaterialAmmonia
ColorColorless
CAS7664-41-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS P Statement Toxic in contact with skin. Causes damage to organs. Toxic if swallowed. Causes serious eye damage. Toxic if inhaled. Causes skin irritation. Highly flammable liquid and vapor.