3-Amino-5-methylpyrazole, 97%
3-Amino-5-methylpyrazole, 97%
3-Amino-5-methylpyrazole, 97%
3-Amino-5-methylpyrazole, 97%
Thermo Scientific Chemicals
3-Amino-5-methylpyrazole, 97%
CAS: 31230-17-8 | C4H7N3 | 97.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR36803025025 gGlass bottle
Catalog number ACR368030250
Price (MYR)
827.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
827.00
EA
Add to cart
Specifications
Chemical Name or Material3-Amino-5-methylpyrazole
Name Note97%
CAS31230-17-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning