Hydrochloric acid, 4N solution in 1,4-Dioxane/water
Hydrochloric acid, 4N solution in 1,4-Dioxane/water
Hydrochloric acid, 4N solution in 1,4-Dioxane/water
Hydrochloric acid, 4N solution in 1,4-Dioxane/water
Thermo Scientific Chemicals
Hydrochloric acid, 4N solution in 1,4-Dioxane/water
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3665200252.5 LGlass bottle
Catalog number ACR366520025
Price (MYR)
3,619.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,619.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 7732-18-5, 123-91-1
Specifications
Chemical Name or MaterialHydrochloric acid
ColorColorless to Yellow
Name Note4N Solution in 1, 4-dioxane/water
CAS7647-01-0
Concentration3.95 to 4.40N