Boron tribromide, 99+%
Boron tribromide, 99+%
Boron tribromide, 99+%
Boron tribromide, 99+%
Thermo Scientific Chemicals
Boron tribromide, 99+%
CAS: 10294-33-4 | BBr3 | 250.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR365211000
Price (MYR)
1,003.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,003.00
EA
Add to cart
Specifications
Boiling Point91.2°C
Chemical Name or MaterialBoron tribromide
Melting Point-46.0°C
Name Note99+%
CAS10294-33-4
View more
C-X bond formation reagent, Lewis acid
RUO – Research Use Only