Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR364320010
1 L
AcroSeal™ Glass Bottle
Catalog number ACR364320010
Price (MYR)
849.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
849.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteOver Molecular Sieves, Stabilized
CAS67-66-3
Health Hazard 1Warning