▸
Organic compounds
▸
Sulfonyl halides
▸
3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number
ACR357080050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-chloro-2-methylbenzenesulfonyl chloride
CAS
80563-86-6
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0