2-Phenylimidazole, 98%
2-Phenylimidazole, 98%
2-Phenylimidazole, 98%
2-Phenylimidazole, 98%
Thermo Scientific Chemicals
2-Phenylimidazole, 98%
CAS: 670-96-2 | C9H8N2 | 144.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR35643025025 gGlass bottle
Catalog number ACR356430250
Price (MYR)
647.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
647.00
EA
Add to cart
Specifications
Chemical Name or Material2-Phenylimidazole
Name Note95%
CAS670-96-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning