1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
Thermo Scientific Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
CAS: 920-66-1 | C3H2F6O | 168.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR354861000100 mLGlass bottle
Catalog number ACR354861000
Price (MYR)
6,010.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,010.00
EA
Add to cart
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexafluoro-2-propanol
CAS920-66-1
CAS Min %99.75
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0