n-Heneicosane, 98%
n-Heneicosane, 98%
n-Heneicosane, 98%
Acros Organics
n-Heneicosane, 98%
CAS: 629-94-7 | C21H44 | 296.58 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3527700101 gGlass bottle
Catalog number ACR352770010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heneicosane
CAS629-94-7
CAS Min %97.5
CAS Max %100.0
Melting Point39°C to 43°C