Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Thermo Scientific Chemicals
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number ACR351425000
Price (MYR)
2,132.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
2,132.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform-d
Density1.5000g/mL
Deuterium (D)99.75% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only