1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
Thermo Scientific Chemicals
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
CAS: 27905-45-9 | C13H7F17O2 | 518.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3486100505 gGlass Bottle
Catalog number ACR348610050
Price (MYR)
759.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
759.00
BOT
Add to cart
Specifications
Chemical Name or Material1H,1H,2H,2H-Heptadecafluorodecyl acrylate
Name Note97%
CAS27905-45-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning