all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
Thermo Scientific Chemicals
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
CAS: 127-47-9 | C22H32O2 | 328.49 g/mol
Have Questions?
Catalog NumberQuantity
ACR346321000100 g
Catalog number ACR346321000
Price (MYR)
1,284.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,284.00
BOT
Add to cart
Specifications
Chemical Name or Materialall-trans-Retinyl acetate
CAS127-47-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.