Phenosafranin
Phenosafranin
Phenosafranin
Thermo Scientific Chemicals
Phenosafranin
CAS: 81-93-6 | C18H15ClN4 | 322.80 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3393400505 gGlass bottle
Catalog number ACR339340050
Price (MYR)
709.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
709.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenosafranin
CAS81-93-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
View more
Indicator
RUO – Research Use Only