Taurodeoxycholic acid, sodium salt hydrate, 97%
Taurodeoxycholic acid, sodium salt hydrate, 97%
Taurodeoxycholic acid, sodium salt hydrate, 97%
Taurodeoxycholic acid, sodium salt hydrate, 97%
Thermo Scientific Chemicals
Taurodeoxycholic acid, sodium salt hydrate, 97%
Taurodeoxycholic acid, sodium salt hydrate, 97%, CAS # 207737-97-1, also known as sodium taurodeoxylate, is a bile acid taurine conjugate of deoxycholic acid, and acts as a detergent. | CAS: 207737-97-1 | C26H44NNaO6S | 521.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3368400505 gGlass bottle
Catalog number ACR336840050
Price (MYR)
1,475.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,475.00
EA
Add to cart
Specifications
Chemical Name or MaterialTaurodeoxycholic acid, sodium salt hydrate
ColorWhite
Name Note97%
CAS207737-97-1
Health Hazard 1
View more
General Description
• Taurodeoxycholic acid, sodium salt hydrate, is a bile acid that is derived from deoxycholic acid
• It is also a conjugate acid of a taurodeoxycholate with detergent activity • It is formed in the liver by conjugating deoxycholate with a taurine group, commonly found in the form of a sodium salt
Applications
• Taurodeoxycholic acid, sodium salt hydrate has been studied to assess the adsorption of bile acids and salts in vegetable fibrous tissue
• It has been used as an anionic detergent important for protein solubilization
• This compound shows cholagogue and choleretic activities
• It has also been used to explore a methodology for determining conjugated and unconjugated bile salts both in serum and jejunal fluids
RUO – Research Use Only
General References:
  1. Schmid, A.; Neumann, H.; Karrasch, T.; Liebisch, G.; Schäffler, A. Bile acid metabolome after an oral lipid tolerance test by liquid chromatography-tandem mass spectrometry (LC-MS/MS). PLoS One. 2016, 11 (2), e0148869.
  2. Tangerman, A.; van Schaik, A.; van der Hoek, E.W. Analysis of conjugated and unconjugated bile acids in serum and jejunal fluid of normal subjects. Clin Chim Acta. 1986 Sep 15;159(2):123-32.