Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Thermo Scientific Chemicals
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR332610010
Price (MYR)
901.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
901.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarboxymethylecellulose, sodium salt
ColorWhite to Beige
Name NoteAverage M.W. 250000 (DS=0.7)
pH6.5 to 8 (1% soln.)
CAS9004-32-4