Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
Acros Organics
Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Plastic bag in resealable tin can
Catalog number ACR332100100
Price (MYR)
-
Quantity:
10 g
Packaging:
Plastic bag in resealable tin can
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium Aluminum hydride
Melting Point125.0°C
Name NoteIn Resealable Tin Cans
CAS16853-85-3
ColorWhite to Gray
View more
Reduction reagent
RUO – Research Use Only