Glipizide, 98+%
Glipizide, 98+%
Glipizide, 98+%
Thermo Scientific Chemicals
Glipizide, 98+%
CAS: 29094-61-9 | C21H27N5O4S | 445.5 g/mol
Have Questions?
Catalog NumberQuantity
ACR3291100101 g
Catalog number ACR329110010
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlipizide
CAS29094-61-9
ColorWhite
EINECS Number249-427-6
Ignition Residue0.4% max.