Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Thermo Scientific Chemicals
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
CAS: 610769-35-2 | C14H30BN3O14 | 475.211 g/mol
Have Questions?
Catalog NumberQuantity
ACR3273300101 L
Catalog number ACR327330010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialTris Borate EDTA buffer
Name NotepH 8.3
CAS610769-35-2
Health Hazard 1Warning
ColorColorless
View more
Chelation/Complexation reagent, pH buffer
RUO – Research Use Only