1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
Thermo Scientific Chemicals
1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3212600101 gGlass Bottle
Catalog number ACR321260010
Price (MYR)
647.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
647.00
EA
Add to cart
Specifications
Chemical Name or Material1, 2-Dichlorobenzene-d4
Density1.3400g/mL
Deuterium (D)97.5% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only