Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
Thermo Scientific Chemicals
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ACR3177300101 Ea.
Catalog number ACR317730010
Price (MYR)
1,672.00
BOT
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (MYR)
1,672.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name Note99.9% (1ea = 10.4gr)
CAS7440-22-4
Melting Point961.0°C
View more
For ultra trace analysis
RUO – Research Use Only