Ammonium tetrafluoroborate, 99%, pure
Ammonium tetrafluoroborate, 99%, pure
Ammonium tetrafluoroborate, 99%, pure
Ammonium tetrafluoroborate, 99%, pure
Thermo Scientific Chemicals
Ammonium tetrafluoroborate, 99%, pure
CAS: 13826-83-0 | BF4H4N | 104.84 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR310961000100 gPlastic bottle
Catalog number ACR310961000
Price (MYR)
444.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
444.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium tetrafluoroborate
Melting Point230.0°C
CAS13826-83-0
ColorColorless
Health Hazard 1GHS Signal Word: Danger