Ethyl 6-bromohexanoate, 98%
Ethyl 6-bromohexanoate, 98%
Ethyl 6-bromohexanoate, 98%
Ethyl 6-bromohexanoate, 98%
Thermo Scientific Chemicals
Ethyl 6-bromohexanoate, 98%
CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR30592025025 gGlass bottle
Catalog number ACR305920250
Price (MYR)
686.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
686.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl 6-bromohexanoate
Name Note98%
CAS25542-62-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.