Purifier™ Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 10in. Opening, International Model
Purifier™ Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 10in. Opening, International Model
Actual product may vary
Thermo Scientific Chemicals
Purifier™ Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 10in. Opening, International Model
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
Have Questions?
Catalog NumberHeight (Metric)IncludesLightingPlug Type
ACR302610050156.7 cmCabinet OnlyFluorescentUK (230 V)
Catalog number ACR302610050
Price (MYR)
742.00
EA
Add to cart
Height (Metric):
156.7 cm
Includes:
Cabinet Only
Lighting:
Fluorescent
Plug Type:
UK (230 V)
Price (MYR)
742.00
EA
Add to cart
MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.
  • Same ECM blower technology as Purifier Logic™; uses CAP (Constant Airflow Profile)™ to maintain precise volume of air as filters load
  • As an additional safety feature, cabinet will continuously display HEPA filter life remaining and cabinet status
  • Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
  • Discharges HEPA-filtered exhaust air directly into lab, or into an exhaust system through optional Canopy Connection
Inclination Technology™ Suite includes:
  • International Model
  • 10° angled sash
  • Patented curved air grille with built-in armrest
  • Fully accessible controls and eye-level mounted color display with MyLogic operating system
Some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. (Refer to nominal width, sash opening, electrical configuration and product specifications for details.)
Specifications
Certifications/ComplianceADA, CE, NSF 49
Chamber MaterialStainless Steel
Noise Level<63 dBA
Plug TypeUK (230 V)
Height (English)61.7 in.
Sash Opening (Metric)25.4 cm
DisplayLCD
Housing MaterialPowder Coated Steel
IncludesCabinet Only
LightingFluorescent
Height (Metric)156.7 cm
Sash Opening (English)10 in.
Width (English)78.3 in.
Width (Metric)198.8 cm
Height (English) Viewing27 in.
Depth (English)31.2 in.
Depth (Metric)79.2 cm
Weight (English)805 lb.
Weight (Metric)365.1 Kg
Voltage230 V
Amperage8 A
Unit SizeEA