(1,2-Dibromoethyl)benzene, 97%
(1,2-Dibromoethyl)benzene, 97%
(1,2-Dibromoethyl)benzene, 97%
(1,2-Dibromoethyl)benzene, 97%
Thermo Scientific Chemicals
(1,2-Dibromoethyl)benzene, 97%
CAS: 93-52-7 | C8H8Br2 | 263.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR299305000500 gPlastic bottle
Catalog number ACR299305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material(1, 2-Dibromoethyl)benzene
Name Note97%
CAS93-52-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger