1,5-Cyclooctadiene, 99%, stabilized
1,5-Cyclooctadiene, 99%, stabilized
1,5-Cyclooctadiene, 99%, stabilized
1,5-Cyclooctadiene, 99%, stabilized
Thermo Scientific Chemicals
1,5-Cyclooctadiene, 99%, stabilized
CAS: 111-78-4 | C8H12 | 108.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2971200252.5 LGlass bottle
Catalog number ACR297120025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 5-Cyclooctadiene
Name NoteStabilized
CAS111-78-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger