Hydrazine hydrate, 55% (Hydrazine, 35%)
Hydrazine hydrate, 55% (Hydrazine, 35%)
Hydrazine hydrate, 55% (Hydrazine, 35%)
Hydrazine hydrate, 55% (Hydrazine, 35%)
Hydrazine hydrate, 55% (Hydrazine, 35%)
Hydrazine hydrate, 55% (Hydrazine, 35%)
Thermo Scientific Chemicals
Hydrazine hydrate, 55% (Hydrazine, 35%)
CAS: 10217-52-4 | H4N2 | 32.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR296815000500 mLGlass bottle
Catalog number ACR296815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point109.4°C
Chemical Name or MaterialHydrazine hydrate
Melting Point-65.0°C
Name Note55% (Hydrazine 35%)
CAS10217-52-4
View more
Reduction reagent
RUO – Research Use Only