2-Nitrophenylselenocyanate, 98%
2-Nitrophenylselenocyanate, 98%
2-Nitrophenylselenocyanate, 98%
2-Nitrophenylselenocyanate, 98%
Thermo Scientific Chemicals
2-Nitrophenylselenocyanate, 98%
CAS: 51694-22-5 | C7H4N2O2Se | 227.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2935800101 gGlass bottle
Catalog number ACR293580010
Price (MYR)
1,108.00
G
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,108.00
G
Add to cart
Specifications
Chemical Name or Material2-Nitrophenylselenocyanate
Name Note99%
CAS51694-22-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger