BOC-L-Valine, 99+%
BOC-L-Valine, 99+%
BOC-L-Valine, 99+%
Thermo Scientific Chemicals
BOC-L-Valine, 99+%
CAS: 13734-41-3 | C10H19NO4 | 217.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR27571025025 gGlass Bottle
Catalog number ACR275710250
Price (MYR)
835.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
835.00
EA
Add to cart
Specifications
Chemical Name or MaterialBOC-L-Valine
Melting Point76°C to 80°C
Name Note99+%
CAS13734-41-3