BOC-L-Methionine, 98%
BOC-L-Methionine, 98%
BOC-L-Methionine, 98%
BOC-L-Methionine, 98%
Thermo Scientific Chemicals
BOC-L-Methionine, 98%
CAS: 2488-15-5 | C10H19NO4S | 249.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR27563025025 gGlass Bottle
Catalog number ACR275630250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBOC-L-Methionine
Melting Point47.0°C to 53.0°C
Name Note99+%
CAS2488-15-5