3-Amino-5-hydroxypyrazole, 98%
3-Amino-5-hydroxypyrazole, 98%
3-Amino-5-hydroxypyrazole, 98%
3-Amino-5-hydroxypyrazole, 98%
Thermo Scientific Chemicals
3-Amino-5-hydroxypyrazole, 98%
CAS: 6126-22-3 | C3H5N3O | 99.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2717200505 gGlass bottle
Catalog number ACR271720050
Price (MYR)
469.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
EA
Add to cart
Specifications
Chemical Name or Material3-Amino-5-hydroxypyrazole
CAS6126-22-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0