3-Amino-4-hydroxybenzoic acid, 98%
3-Amino-4-hydroxybenzoic acid, 98%
3-Amino-4-hydroxybenzoic acid, 98%
3-Amino-4-hydroxybenzoic acid, 98%
Thermo Scientific Chemicals
3-Amino-4-hydroxybenzoic acid, 98%
CAS: 1571-72-8 | C7H7NO3 | 153.14 g/mol
Have Questions?
Catalog NumberQuantity
ACR2714900505 g
Catalog number ACR271490050
Price (MYR)
437.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
437.00
EA
Add to cart
Specifications
Chemical Name or Material3-Amino-4-hydroxybenzoic acid
Name Note98%
CAS1571-72-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning