Tetramethylsilane, 99%
Tetramethylsilane, 99%
Tetramethylsilane, 99%
Tetramethylsilane, 99%
Thermo Scientific Chemicals
Tetramethylsilane, 99%
CAS: 75-76-3 | C4H12Si | 88.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR268865000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylsilane
Name Note99%
CAS75-76-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Extremely flammable liquid and vapor.