Chloroform, for HPLC, stabilized with amylene
Chloroform, for HPLC, stabilized with amylene
Chloroform, for HPLC, stabilized with amylene
Chloroform, for HPLC, stabilized with amylene
Thermo Scientific Chemicals
Chloroform, for HPLC, stabilized with amylene
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2683200101 LGlass Bottle
Catalog number ACR268320010
Price (MYR)
512.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
512.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 280nm, 0.01 max. at 270nm, 0.075 max. at 260nm, 0.5 max. at 250nm, 1 max. at 245nm
CAS67-66-3
View more
For HPLC
RUO – Research Use Only