Ytterbium(III) oxide, 99.99%, (trace metal basis)
Ytterbium(III) oxide, 99.99%, (trace metal basis)
Ytterbium(III) oxide, 99.99%, (trace metal basis)
Ytterbium(III) oxide, 99.99%, (trace metal basis)
Thermo Scientific Chemicals
Ytterbium(III) oxide, 99.99%, (trace metal basis)
CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol
Have Questions?
Catalog number ACR264300100
Price (MYR)
1,239.00
Price (MYR)
1,239.00
EA
Add to cart
Specifications
Chemical Name or MaterialYtterbium(III) oxide
Melting Point2346.0°C
Name Note99%
CAS1314-37-0
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only